Merck
  • [Distribution of puerarin contained in Zige freeze-dried powder injection in rat liver and kidney].
[Distribution of puerarin contained in Zige freeze-dried powder injection in rat liver and kidney].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-09-22)
Qin Wang, Mao Xing, Lijuan Deng, Yongsheng Wang, Kipaytul Mukaram, Xiaoyu Xu
ABSTRACT
To investigate the distribution process of puerarin contained in Zige freeze-dried powder injection in rat liver and kidney and the safety of Zige freeze-dried powder injection. Rats were divided into the Zige freeze-dried powder injection group and the puerarin freeze-dried powder injection control group randomly. The liver and kidney samples were collected at 5, 10, 20, 30, 45, 60 and 120 min after intravenous administration of puerarin (26.7 microg x g(-1)) through caudal vein and detected by HPLC. The concentration of puerarin in kidney reached the max value of 58.12 microg x g(-1) for the Zige freeze-dried powder injection group and 71.28 microg x g(-1) for the Puerarin freeze-dried powder injection group. The value of AUC(0-2h) was 26.24 microg x h x g(-1) for the Zige freeze-dried powder injection group and 35.24 microg x h x g(-1) for the puerarin freeze-dried powder injection group, MRT(0-2h) was 0. 39 h for the Zige freeze-dried powder injection group and 0. 42 h for the puerarin freeze-dried powder injection control group. Compared with the control group, the Zige freeze-dried powder injection group showed a significant decrease in Cmax and AUC(0-2h) (P < 0.05), with no notable difference in peak time tmax and MRT(0-2h). The two groups showed no obvious difference in tmax Cmax, AUC(0-2h) and MRT(0-2h) of puerarin in rat kidney. Compared with Zige freeze-dried powder injection, Zige freeze-dried powder injection can reduce the distribution of puerarin in rat kidney, with no obvious change in the elimination of puerarin in rat kidney. It also showed no significant change in distribution and elimination in liver. This indicates that Zige freeze-dried powder injection is safer to kidney than puerarin freeze-dried powder injection.
Sigma-Aldrich
Puerarin, ≥98.0% (HPLC)
Supelco
Puerarin, analytical standard