Merck
  • Novel formulations of dipyridamole with microenvironmental pH-modifiers for improved dissolution and bioavailability under hypochlorhydria.
Novel formulations of dipyridamole with microenvironmental pH-modifiers for improved dissolution and bioavailability under hypochlorhydria.
International journal of pharmaceutics (2012-05-31)
Chika Taniguchi, Ryo Inoue, Yohei Kawabata, Kazuhiro Yamashita, Koichi Wada, Yukinori Yamauchi, Shizuo Yamada, Satomi Onoue
ABSTRACT
This study was undertaken to develop new dipyridamole (DP) formulations with acidic microenvironmental pH-modifiers for improving dissolution and absorption under hypochlorhydric conditions. Dipyridamole granules (DPG) with ten acidic pH-modifiers were prepared with conventional wet granulation, and their manufacturability, stability and dissolution behavior were characterized. Pharmacokinetic profiling of the optimized DPG with acid was carried out in omeprazole-treated rats as a hypochlorhydric model. On the basis of the manufacturability, stability and dissolution behavior of new DPG formulations, p-toluenesulfonic acid (TS) was found to be a suitable acidic pH-modifier for DPG formulation. Although DPG showed pH-dependent dissolution behavior, DPG with TS exhibited a high rate and extent of dissolution in both acidic and neutral media. After oral administration of DPG (10mg DP/kg) in omeprazole-treated hypochlorhydric rats, there was ca. 40% reduction of the area under the curve of plasma concentration vs. time from zero to 3h (AUC(0-3)) for DPG compared with that in normal rats. However, AUC(0-3) for DPG/TS under hypochlorhydria was almost identical to that of DPG in normal rats. From these findings, the addition of TS as a microenvironmental pH-modifier in DP formulation might be beneficial in expanding the therapeutic potential of DP in hypochlorhydric patients.
Sigma-Aldrich
Silver p-toluenesulfonate, ≥99%
Sigma-Aldrich
p-Toluenesulfonic acid monohydrate, ReagentPlus®, ≥98%
Sigma-Aldrich
p-Toluenesulfonic acid monohydrate, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium p-toluenesulfonate, 95%