Merck
  • Precipitation of dispersed silver particles using acetone as reducing agent.
Precipitation of dispersed silver particles using acetone as reducing agent.
Journal of colloid and interface science (2010-12-08)
Ionel Halaciuga, Sylas Laplante, Dan V Goia
ABSTRACT
A novel environmentally friendly solution-based method for preparing dispersed silver particles is described. The simple and convenient approach consists in heating silver oxide particles dispersed in a highly alkaline water/acetone mixture. The data presented clearly show that acetone reduces completely and rapidly Ag(2)O particles to metallic silver at 60°C. A mechanism explaining the provenance of the electrons responsible for the reduction of silver is proposed.
Sigma-Aldrich
Silver oxide, predominantly silver(II) oxide
Sigma-Aldrich
Silver(I) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Silver(I) oxide, purum p.a., ≥99.0% (AT)
Sigma-Aldrich
Silver(I) oxide, ReagentPlus®, 99%