Merck
  • 2-chlorophenyl zinc bromide: a convenient nucleophile for the mannich-related multicomponent synthesis of clopidogrel and ticlopidine.
2-chlorophenyl zinc bromide: a convenient nucleophile for the mannich-related multicomponent synthesis of clopidogrel and ticlopidine.
Molecules (Basel, Switzerland) (2010-11-13)
Isabelle Aillaud, Caroline Haurena, Erwan Le Gall, Thierry Martens, Gino Ricci
ABSTRACT
A methodological study devoted to the Mannich-like multicomponent synthesis of the antiplatelet agent (±)‑clopidogrel (7) and the ethyl ester analogue 6 is described. The process involves the formation of 2-chlorophenyl zinc bromide (2) and its subsequent reaction with an alkyl glyoxylate and 4,5,6,7-tetrahydrothieno[3,2-c]pyridine (3). We demonstrate that the organozinc reagent 2 also constitutes a very convenient nucleophile for the multicomponent synthesis of the benzylamine core of ticlopidine (9).
Sigma-Aldrich
Zinc bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc bromide, 99.999% trace metals basis
Sigma-Aldrich
Zinc bromide, puriss., anhydrous, ≥98%