Merck
  • Reaction of ADP with amino acid methyl esters mediated by trimethylsilyl chloride.
Reaction of ADP with amino acid methyl esters mediated by trimethylsilyl chloride.
Chemical communications (Cambridge, England) (2003-03-04)
Hua Fu, Bo Han, Yu-Fen Zhao
ABSTRACT
Reaction of ADP with amino acid methyl esters mediated by trimethylsilyl chloride in pyridine produced adenosine 5'-phosphoramidates in good yields under mild conditions, it is interesting that nucleophilic attack of amino acid methyl esters only occurred on alpha-phosphorus of ADP.
Sigma-Aldrich
L-Phenylalanine methyl ester hydrochloride
Sigma-Aldrich
L-Phenylalanine methyl ester hydrochloride, 98%
Sigma-Aldrich
D-Phenylalanine methyl ester hydrochloride, 98%