Merck
Molecular mechanism of staurosporine-induced apoptosis in osteoblasts.
Pharmacological research (2000-09-16)
H J Chae, J S Kang, J O Byun, K S Han, D U Kim, S M Oh, H M Kim, S W Chae, H R Kim
ABSTRACT
Staurosporine, a microbial alkaloid, is a strong inhibitor of protein kinases. We induced apoptosis in murine osteoblast MC3T3E-1 cells by exposure to the staurosporine. Staurosporine transiently increased the phosphotransferase activity of c-Jun N-terminal kinase-1 (JNK1), which in turn may activate the transcriptional activity of activating protein-1 (AP-1). We then prepared extracts from staurosporine-treated MC3T3E-1 cells and monitored the cleavage of acetyl-YVAD-AMC and acetyl-DEVD-AMC, fluorogenic substrates of caspase-1-like and caspase-3-like proteases, respectively. Staurosporine caused a significant increase in the proteolytic activity of caspase-3-like proteases, but not in the activity of caspase-1-like proteases. Furthermore, staurosporine increased the transcriptional activity of nuclear factor- kappa B (NF- kappa B). These data suggest that staurosporine-induced apoptosis in osteoblasts may occur via activation of JNK1, caspase-3-like proteases, and transcriptional factors including AP-1 and NF- kappa B.
Sigma-Aldrich
Staurosporine from Streptomyces sp., ≥98% (HPLC), film
Sigma-Aldrich
Staurosporine from Streptomyces sp., ≥95% (HPLC)