Merck
  • Neuropeptide Y regulates cholesterol uptake and efflux in macrophages and promotes foam cell formation.
Neuropeptide Y regulates cholesterol uptake and efflux in macrophages and promotes foam cell formation.
Journal of cellular and molecular medicine (2022-09-30)
Yu Cai, Zhengchao Wang, Lun Li, Li He, Xinying Wu, Mingjing Zhang, Pengfei Zhu
ABSTRACT
The dysregulation of lipid metabolic pathways (cholesterol uptake and efflux) in macrophages results in the formation of lipid-dense macrophages, named foam cells, that participate in plaque formation. NPY binding to NPY receptors in macrophages can modulate cell functions and affect the process of atherosclerotic plaques. The present study aimed to determine whether NPY affects the formation of macrophage-derived foam cells and its underlying mechanisms in macrophages. THP-1-derived macrophages were incubated with oxidized low-density lipoprotein (ox-LDL) and treated with different concentrations of NPY. We analysed the relative levels of proteins related to cholesterol uptake and efflux. We found that NPY effectively increased cholesterol uptake and intracellular cholesterol content via the Y1 and Y5 receptors, and this effect was blocked by Y1 and Y5 antagonists. Mechanistically, NPY enhanced the expression of SRA and CD36 via the PKC/PPARγ pathways, promoting macrophage cholesterol uptake. Moreover, NPY significantly decreased cholesterol efflux to the extracellular cholesterol acceptors ApoA1 and HDL in macrophages. NPY mediated decreases in ABCA1, ABCG1 and SR-BI expression through the inhibition of the JAK/STAT3 pathways. Our results suggest that NPY binding to the Y1 and Y5 receptors enhances foam cell formation by regulating cholesterol uptake and efflux in macrophages.
Sigma-Aldrich
Anti-phospho-JAK2 (pTyr1007) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-PKC α antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-PKC α (pTyr657) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-JAK2 antibody produced in rabbit, affinity isolated antibody