Merck
  • Gasdermin D-mediated pyroptosis suppresses liver regeneration after 70% partial hepatectomy.
Gasdermin D-mediated pyroptosis suppresses liver regeneration after 70% partial hepatectomy.
Hepatology communications (2022-05-06)
Xingyu Lv, Jiang Chen, Jiayan He, Lidan Hou, Yiyue Ren, Xiaoyun Shen, Yifan Wang, Tong Ji, Xiujun Cai
ABSTRACT
Pyroptosis is a kind of programmed cell death primarily mediated by gasdermin D (GSDMD) and shown to regulate multiple diseases. However, its contribution to liver regeneration, a fine-tuned tissue repair process mediated primarily by hepatocytes after mass loss, remains unclear. Herein, we found that caspase-11/GSDMD-mediated pyroptosis was activated in regenerating liver after 70% partial hepatectomy. Impeding pyroptosis by deleting GSDMD significantly reduced liver injury and accelerated liver regeneration. Mechanistically, GSDMD deficiency up-regulates the activation of hepatocyte growth factor/c-Met and epidermal growth factor receptor mitogenic pathways at the initiation phase. Moreover, activin A and glypican 3 (GPC3), two terminators of liver regeneration, were inhibited when GSDMD was absent. In vitro study suggested the expressions of activin A and GPC3 were induced by interleukin (IL)-1β and IL-18, whose maturations were regulated by GSDMD-mediated pyroptosis. Similarly, pharmacologically inhibiting GSDMD recapitulates these phenomena. Conclusion: This study characterizes the role of GSDMD-mediated pyroptosis in liver regeneration and lays the foundation for enhancing liver restoration by targeting GSDMD in liver patients with impaired regenerative capacity.
Sigma-Aldrich
Anti-BrdU Antibody, clone PRB-1, biotin conjugated, clone PRB-1, Chemicon®, from mouse
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid