Merck
  • Activity-regulated E3 ubiquitin ligase TRIM47 modulates excitatory synapse development.
Activity-regulated E3 ubiquitin ligase TRIM47 modulates excitatory synapse development.
Frontiers in molecular neuroscience (2022-10-11)
Gourav Sharma, Sourav Banerjee
ABSTRACT
The Ubiquitin Proteasome System (UPS) has been shown to regulate neuronal development and synapse formation. Activity-dependent regulation of E3 ligase, a component of the UPS that targets specific proteins for proteasome-mediated degradation, is emerging as a pivotal player for the establishment of functional synapses. Here, we identified TRIM47 as a developmentally regulated E3 ligase that is expressed in rat hippocampus during the temporal window of synapse formation. We have demonstrated that the expression of TRIM47 is regulated by the glutamate-induced synaptic activity of hippocampal neurons in culture. In addition, the activity-dependent enhancement of TRIM47 expression is recapitulated following the object location test, a hippocampus-dependent spatial memory paradigm. We observed that this enhancement of TRIM47 expression requires NMDA receptor activation. The knockdown of TRIM47 leads to an enhancement of spine density without affecting dendritic complexity. Furthermore, we observed an increase in excitatory synapse development upon loss of TRIM47 function. Comprehensively, our study identified an activity-regulated E3 ligase that drives excitatory synapse formation in hippocampal neurons.
Sigma-Aldrich
Anti-c-Fos (Ab-1) Mouse mAb (2G9C3), liquid, clone 2G9C3, Calbiochem®
Sigma-Aldrich
Anti-Synapsin I Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-β-Tubulin III (neuronal) antibody, Mouse monoclonal, ~1.0 mg/mL, clone 2G10, purified from hybridoma cell culture