Merck
  • A protocol for CRISPR-mediated activation and repression of human endogenous retroviruses in human pluripotent stem cells.
A protocol for CRISPR-mediated activation and repression of human endogenous retroviruses in human pluripotent stem cells.
STAR protocols (2022-04-26)
Vidya Padmanabhan Nair, Jens Mayer, Michelle Vincendeau
ABSTRACT
Human endogenous retroviruses (HERVs) comprise many regulatory elements and can regulate host gene activity at different expression levels via multiple mechanisms. Here, we introduce a step-by-step protocol to activate or repress transcription of HERV-K(HML-2) elements using the CRISPRa and CRISPRi technologies in human embryonic stem cells. This protocol can help deciphering the functional role of HERV-K(HML-2) elements in critical biological processes. The protocol may easily be adapted to other cell lines and HERV groups with relatively low sequence heterogeneity. For complete details on the use and execution of this protocol, please refer to Padmanabhan Nair et al. (2021).
Sigma-Aldrich
LB Broth with agar (Luria low salt), Powder microbial growth medium
Sigma-Aldrich
LB Broth (Luria low salt), Powder microbial growth medium
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%