Merck
Lung Section Staining and Microscopy.
Bio-protocol (2017-11-25)
Xiaofeng Zhou, Bethany B Moore
ABSTRACT
Our protocol describes immunofluorescent staining, hematoxylin and eosin staining and Masson's trichrome staining on lung sections.
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Iron(III) chloride, reagent grade, 97%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%