Merck
  • Exploiting the lipoic acid structure in the search for novel multitarget ligands against Alzheimer's disease.
Exploiting the lipoic acid structure in the search for novel multitarget ligands against Alzheimer's disease.
European journal of medicinal chemistry (2011-09-20)
Michela Rosini, Elena Simoni, Manuela Bartolini, Andrea Tarozzi, Riccardo Matera, Andrea Milelli, Patrizia Hrelia, Vincenza Andrisano, Maria Laura Bolognesi, Carlo Melchiorre
ABSTRACT
Lipoic acid (LA) is a natural antioxidant. Its structure was previously combined with that of the acetylcholinesterase inhibitor tacrine to give lipocrine (1), a lead compound multitargeted against Alzheimer's disease (AD). Herein, we further explore LA as a privileged structure for developing multimodal compounds to investigate AD. First, we studied the effect of LA chirality by evaluating the cholinesterase profile of 1's enantiomers. Then, a new series of LA hybrids was designed and synthesized by combining racemic LA with motifs of other known anticholinesterase agents (rivastigmine and memoquin). This afforded 4, which represents a step forward in the search for balanced anticholinesterase and antioxidant capacities.
Sigma-Aldrich
(±)-α-Lipoic acid, synthetic, ≥99% (titration), powder
Sigma-Aldrich
(±)-α-Lipoic acid, ≥98.0%
Sigma-Aldrich
(±)-α-Lipoic acid, suitable for cell culture, BioReagent, ≥99%