Merck
  • H3K27M in Gliomas Causes a One-Step Decrease in H3K27 Methylation and Reduced Spreading within the Constraints of H3K36 Methylation.
H3K27M in Gliomas Causes a One-Step Decrease in H3K27 Methylation and Reduced Spreading within the Constraints of H3K36 Methylation.
Cell reports (2020-11-19)
Ashot S Harutyunyan, Haifen Chen, Tianyuan Lu, Cynthia Horth, Hamid Nikbakht, Brian Krug, Caterina Russo, Eric Bareke, Dylan M Marchione, Mariel Coradin, Benjamin A Garcia, Nada Jabado, Jacek Majewski
ABSTRACT
The discovery of H3K27M mutations in pediatric gliomas marked a new chapter in cancer epigenomics. Numerous studies have investigated the effect of this mutation on H3K27 trimethylation, but only recently have we started to realize its additional effects on the epigenome. Here, we use isogenic glioma H3K27M+/- cell lines to investigate H3K27 methylation and its interaction with H3K36 and H3K9 modifications. We describe a "step down" effect of H3K27M on the distribution of H3K27 methylation: me3 is reduced to me2, me2 is reduced to me1, whereas H3K36me2/3 delineates the boundaries for the spread of H3K27me marks. We also observe a replacement of H3K27me2/3 silencing by H3K9me3. Using a computational simulation, we explain our observations by reduced effectiveness of PRC2 and constraints imposed on the deposition of H3K27me by antagonistic H3K36 modifications. Our work further elucidates the effects of H3K27M in gliomas as well as the general principles of deposition in H3K27 methylation.
Sigma-Aldrich
Anti-trimethyl Histone H3 (Lys27) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-WHSC1/NSD2 Antibody, clone 29D1, clone 29D1, from mouse
Sigma-Aldrich
Anti-Histone H3.3 Antibody, K27M mutant, from rabbit, purified by affinity chromatography