Merck
  • Impact of glycosylation and detergent-resistant membranes on the function of intestinal sucrase-isomaltase.
Impact of glycosylation and detergent-resistant membranes on the function of intestinal sucrase-isomaltase.
Biological chemistry (2009-05-12)
Gabi Wetzel, Martin Heine, Arndt Rohwedder, Hassan Y Naim
ABSTRACT
Sucrase-isomaltase (SI) is a highly N- and O-glycosylated intestinal brush border membrane protein. SI is sorted with high fidelity to the apical membrane via O-linked glycans that mediate its association with lipid rafts or detergent-resistant membranes (DRMs). Here, we show that N- and O-glycosylation and DRMs are implicated in the regulation of the function of SI in intestinal Caco-2 cells. The activities of sucrase (SUC) and isomaltase (IM) increase substantially in DRMs when N- and O-glycosylation patterns are intact. Disruption of DRMs by solubilization with Triton X-100 at 37 degrees C substantially reduces the activities of SUC and IM. Furthermore, modulation of O-glycosylation by benzyl-2-acetamido-2-deoxy-alpha-d-galactopyranoside and N-glycosylation by deoxymannojirimycin is linked to a decreased capacity of SI to associate with DRMs, with a subsequent reduction of the enzymatic activities of SUC and IM. This is the first report that reveals a direct role of N- and O-glycans in association with DRMs in regulating the function of a membrane glycoprotein.
Sigma-Aldrich
Benzyl 2-acetamido-2-deoxy-α-D-galactopyranoside, O-glycosylation inhibitor, ≥97% (TLC)