Merck
  • Barrier-to-autointegration factor (BAF) involvement in prelamin A-related chromatin organization changes.
Barrier-to-autointegration factor (BAF) involvement in prelamin A-related chromatin organization changes.
Oncotarget (2015-12-25)
Manuela Loi, Vittoria Cenni, Serena Duchi, Stefano Squarzoni, Carlos Lopez-Otin, Roland Foisner, Giovanna Lattanzi, Cristina Capanni
ABSTRACT
Chromatin disorganization is one of the major alterations linked to prelamin A processing impairment. In this study we demonstrate that BAF is necessary to modulate prelamin A effects on chromatin structure. We show that when prelamin A and BAF cannot properly interact no prelamin A-dependent effects on chromatin occur; similar to what is observed in human Nestor Guillermo Progeria Syndrome cells harboring a BAF mutation, in HEK293 cells expressing a BAF mutant unable to bind prelamin A, or in siRNA mediated BAF-depleted HEK293 cells expressing prelamin A. BAF is necessary to induce histone trimethyl-H3K9 as well as HP1-alpha and LAP2-alpha nuclear relocalization in response to prelamin A accumulation. These findings are enforced by electron microscopy evaluations showing how the prelamin A-BAF interaction governs overall chromatin organization. Finally, we demonstrate that the LAP2-alpha nuclear localization defect observed in HGPS cells involves the progerin-BAF interaction, thus establishing a functional link between BAF and prelamin A pathological forms.
Sigma-Aldrich
Anti-Progerin Antibody, clone 13A4, clone 13A4, Upstate®, from mouse