Merck
  • Inhibition of endothelial ERK signalling by Smad1/5 is essential for haematopoietic stem cell emergence.
Inhibition of endothelial ERK signalling by Smad1/5 is essential for haematopoietic stem cell emergence.
Nature communications (2014-03-13)
Chunxia Zhang, Junhua Lv, Qiuping He, Sifeng Wang, Ya Gao, Anming Meng, Xiao Yang, Feng Liu
ABSTRACT
The earliest HSCs are derived from haemogenic endothelium via endothelial-to-haematopoietic transition during vertebrate embryogenesis; however, the underlying mechanism is largely unclear. Here we show that interplay of Smad1/5 and ERK signalling is essential for haemogenic endothelium-based HSC emergence. Smad1/5 directly inhibits erk expression through recruiting HDAC1 to and inducing de-acetylation of the erk promoter in endothelial cells. Over-activated ERK signalling conferred by inhibition of Smad1/5 promotes the arterial endothelial cell fate and constitutively strengthens the tight junction between endothelial cells, thereby repressing the specification of haemogenic endothelium and the following endothelial-to-haematopoietic transition process. These findings provide new insights into the in vitro generation of transplantable HSCs for potential clinical applications.
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys9) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-Klf2 Antibody, serum, from rabbit
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks