Merck
  • Abdominal Aortic Aneurysms Growth Is Associated With High Concentrations of Plasma Proteins in the Intraluminal Thrombus and Diseased Arterial Tissue.
Abdominal Aortic Aneurysms Growth Is Associated With High Concentrations of Plasma Proteins in the Intraluminal Thrombus and Diseased Arterial Tissue.
Arteriosclerosis, thrombosis, and vascular biology (2018-07-14)
Carsten Behr Andersen, Jes S Lindholt, Sigitas Urbonavicius, Ulrich Halekoh, Pia Søndergaard Jensen, Jane Stubbe, Lars Melholt Rasmussen, Hans Christian Beck
ABSTRACT
Objective- Porosity of the intraluminal thrombus (ILT) is believed to convey biologically active components from the bloodstream toward the aneurismal wall. Accumulation of molecules in the abdominal aortic aneurysmatic tissue may influence vascular protein turnover and regulate abdominal aortic aneurysm growth. We sought to identify proteins with concentrations in the ILT and the abdominal aortic aneurysm wall which associate with aneurysmal expansion rate. Approach and Results- Proteomic analysis by liquid chromatography tandem-mass spectrometry of separated wall and ILT samples was correlated with preoperative aneurysmal growth rate in 24 individuals operated electively for infrarenal abdominal aortic aneurysm. The median preoperative growth rate was 3.8 mm/y (interquartile range, 3) and the mean observational time was 3.3±1.7 years. Plasma components dominated the group of proteins with tissue concentrations, which correlate positively with growth rates ( P<0.001, Fisher exact test, both in the ILT and the wall). In contrast, in the wall and thrombus samples, ECM (extracellular matrix) proteins were significantly more prevalent in the group of proteins with negative correlations to growth rates ( P<0.05, Fisher exact test). Similarly, a long series of proteins, related to cellular functions correlated negatively to growth rates. Conclusions- When the preoperative aneurysmatic growth rate has been high, the concentration of many plasma proteins residing in the ILT and the aneurysmatic tissue is also high, compatible with the hypothesis of increased tissue porosity and accumulation of plasma components as a driver of aneurysm expansion. Moreover, many matrix and cellular proteins which are found in high concentrations in slower-growing aneurysms provides new knowledge about potential treatment targets.
Sigma-Aldrich
Anti-SERPINA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-AFM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-XRCC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution