Merck
  • The antioxidant vitamin E as a membrane raft modulator: Tocopherols do not abolish lipid domains.
The antioxidant vitamin E as a membrane raft modulator: Tocopherols do not abolish lipid domains.
Biochimica et biophysica acta. Biomembranes (2020-01-19)
Mitchell DiPasquale, Michael H L Nguyen, Brett W Rickeard, Nicole Cesca, Christopher Tannous, Stuart R Castillo, John Katsaras, Elizabeth G Kelley, Frederick A Heberle, Drew Marquardt
ABSTRACT
The antioxidant vitamin E is a commonly used vitamin supplement. Although the multi-billion dollar vitamin and nutritional supplement industry encourages the use of vitamin E, there is very little evidence supporting its actual health benefits. Moreover, vitamin E is now marketed as a lipid raft destabilizing anti-cancer agent, in addition to its antioxidant behaviour. Here, we studied the influence of vitamin E and some of its vitamers on membrane raft stability using phase separating unilamellar lipid vesicles in conjunction with small-angle scattering techniques and fluorescence microscopy. We find that lipid phase behaviour remains unperturbed well beyond physiological concentrations of vitamin E (up to a mole fraction of 0.10). Our results are consistent with a proposed line active role of vitamin E at the domain boundary. We discuss the implications of these findings as they pertain to lipid raft modification in native membranes, and propose a new hypothesis for the antioxidant mechanism of vitamin E.
Product Description
CellCrown inserts, 6 well plate inserts with 1.0 μm polycarbonate filter, sterile
Avanti
16:0 Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
16:0 Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
16:0 PC-d62, Avanti Research - A Croda Brand 860355C
Avanti
16:0 PC-d62, Avanti Research - A Croda Brand 860355P, powder
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand