Merck
  • Identification of a p53 target, CD137L, that mediates growth suppression and immune response of osteosarcoma cells.
Identification of a p53 target, CD137L, that mediates growth suppression and immune response of osteosarcoma cells.
Scientific reports (2017-09-08)
Yusuke Tsuda, Chizu Tanikawa, Takafumi Miyamoto, Makoto Hirata, Varalee Yodsurang, Yao-Zhong Zhang, Seiya Imoto, Rui Yamaguchi, Satoru Miyano, Hiroshi Takayanagi, Hirotaka Kawano, Hidewaki Nakagawa, Sakae Tanaka, Koichi Matsuda
ABSTRACT
p53 encodes a transcription factor that transactivates downstream target genes involved in tumour suppression. Although osteosarcoma frequently has p53 mutations, the role of p53 in osteosarcomagenesis is not fully understood. To explore p53-target genes comprehensively in calvarial bone and find out novel druggable p53 target genes for osteosarcoma, we performed RNA sequencing using the calvarial bone and 23 other tissues from p53 +/+ and p53 -/- mice after radiation exposure. Of 23,813 genes, 69 genes were induced more than two-fold in irradiated p53 +/+ calvarial bone, and 127 genes were repressed. Pathway analysis of the p53-induced genes showed that genes associated with cytokine-cytokine receptor interactions were enriched. Three genes, CD137L, CDC42 binding protein kinase gamma and Follistatin, were identified as novel direct p53 target genes that exhibited growth-suppressive effects on osteosarcoma cell lines. Of the three genes, costimulatory molecule Cd137l was induced only in calvarial bone among the 24 tissues tested. CD137L-expressing cells exhibited growth-suppressive effects in vivo. In addition, recombinant Fc-fusion Cd137l protein activated the immune response in vitro and suppressed osteosarcoma cell growth in vivo. We clarified the role of CD137L in osteosarcomagenesis and its potential therapeutic application. Our transcriptome analysis also indicated the regulation of the immune response through p53.
Sigma-Aldrich
Anti-p21WAF1 Antibody, clone EA10, clone EA10, from mouse
Sigma-Aldrich
EZ-Magna ChIP® G - Chromatin Immunoprecipitation Kit, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic G beads. Control primers included.