Merck
  • Chronic restraint stress induces hippocampal memory deficits by impairing insulin signaling.
Chronic restraint stress induces hippocampal memory deficits by impairing insulin signaling.
Molecular brain (2018-07-05)
Hanwoong Woo, Caroline Jeeyeon Hong, Seonghee Jung, Seongwon Choe, Seong-Woon Yu
ABSTRACT
Chronic stress is a psychologically significant factor that impairs learning and memory in the hippocampus. Insulin signaling is important for the development and cognitive function of the hippocampus. However, the relation between chronic stress and insulin signaling at the molecular level is poorly understood. Here, we show that chronic stress impairs insulin signaling in vitro and in vivo, and thereby induces deficits in hippocampal spatial working memory and neurobehavior. Corticosterone treatment of mouse hippocampal neurons in vitro caused neurotoxicity with an increase in the markers of autophagy but not apoptosis. Corticosterone treatment impaired insulin signaling from early time points. As an in vivo model of stress, mice were subjected to chronic restraint stress. The chronic restraint stress group showed downregulated insulin signaling and suffered deficits in spatial working memory and nesting behavior. Intranasal insulin delivery restored insulin signaling and rescued hippocampal deficits. Our data suggest that psychological stress impairs insulin signaling and results in hippocampal deficits, and these effects can be prevented by intranasal insulin delivery.
Sigma-Aldrich
Anti-p62/SQSTM1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)