Merck
Mechanisms of nutritional and hormonal regulation of lipogenesis.
EMBO reports (2001-04-18)
S Kersten
ABSTRACT
Fat build-up is determined by the balance between lipogenesis and lipolysis/fatty acid oxidation. In the past few years, our understanding of the nutritional, hormonal and particularly transcriptional regulation of lipogenesis has expanded greatly. Lipogenesis is stimulated by a high carbohydrate diet, whereas it is inhibited by polyunsaturated fatty acids and by fasting. These effects are partly mediated by hormones, which inhibit (growth hormone, leptin) or stimulate (insulin) lipogenesis. Recent research has established that sterol regulatory element binding protein-1 is a critical intermediate in the pro- or anti-lipogenic action of several hormones and nutrients. Another transcription factor implicated in lipogenesis is the peroxisome proliferator activated receptor gamma. Both transcription factors are attractive targets for pharmaceutical intervention of disorders such as hypertriglyceridemia and obesity.
Sigma-Aldrich
PPAR, α human, recombinant, expressed in E. coli, ≥75% (SDS-PAGE)
Sigma-Aldrich
PPARα, ligand binding domain (170-468) human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
PPARα isotype, GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE)