
Zenix®-C SEC-150 (3 µm) HPLC Columns

Synonym(s):
Zenix®-C SEC-150 Gel Filtration Column, 3 μm
Products
Product Category
Brand
Manufacturer
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing