
Vanadium(V) oxide

Synonym(s):
Divanadium pentaoxide, Divanadium pentoxide, Pentaoxodivanadium, Vandia, Vanadium(V) oxide
Linear Formula:
V2O5
CAS Number:
Molecular Weight:
181.88
Products
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Chemical Composition
Quality Segment
Application
Reagent Type
Core
Showing 1-4 of 4