
Tetrakis(triphenylphosphine)nickel(0)

Synonym(s):
Nickel-tetrakis(triphenylphosphine)
Linear Formula:
[(C6H5)3P]4Ni
CAS Number:
Molecular Weight:
1107.84
Products
Brand
Formula Weight
Chemical Composition
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing