
Tebufenpyrad

Synonym(s):
4-Chloro-N-(4-tert-butylbenzyl)-3-ethyl-1-methyl-1H-pyrazole-5-carboxamide
Empirical Formula (Hill Notation):
C18H24ClN3O
CAS Number:
Molecular Weight:
333.86
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing