
Suprofen

Synonym(s):
α-Methyl-p-(2-thenoyl)phenylacetic acid
Empirical Formula (Hill Notation):
C14H12O3S
CAS Number:
Molecular Weight:
260.31
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing