Skip to Content
Merck
SPME StableFlex™ Fiber Assortment Kit needle size 23 ga, Autosampler, pk of 4

SPME Fiber Assortment Kit

SPME StableFlex™ Fiber Assortment Kit needle size 23 ga, Autosampler, pk of 4

Products

Brand
Sustainability
Feature
Length (cm)
Application
Color
df (µm)
Package Size
Size
Gauge
Technique
Showing 1-11 of 11
Compare
Product Number
Description
Pricing