Skip to Content
Merck
Sodium pyruvate anhydrous, free-flowing, Redi-Dri™, ReagentPlus®, ≥99%

Sodium pyruvate

Sodium pyruvate anhydrous, free-flowing, Redi-Dri™, ReagentPlus®, ≥99%
Synonym(s):
α-Ketopropionic acid sodium salt, 2-Oxopropanoic acid sodium salt, Pyruvic acid sodium salt
Linear Formula:
CH3COCOONa
CAS Number:
Molecular Weight:
110.04
EC Number:

Products

Product Category
Brand
Biological Source
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Research Area
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Technique