
Rubidium carbonate

Synonym(s):
Carbonic acid dirubidium, Dirubidium carboxide, Rubidium carbonate
Linear Formula:
Rb2CO3
CAS Number:
Molecular Weight:
230.94
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-3 of 3
Compare
Product Number
Description
Pricing