
Rink amide 4-methylbenzhydrylamine, polymer-bound

Synonym(s):
4-(2′,4′-Dimethoxyphenyl-Fmoc-aminmethyl)-phenoxyacetamido-methylbenzhydryl amine resin, Rink amide MBHA resin
Products
Brand
Physical Form
Quality Segment
Functional Group
Application
Cleavage Condition
Linker
Chain Type
Reaction Type
Showing 1-2 of 2