
Raloxifene hydrochloride for peak identification

Synonym(s):
Raloxifene hydrochloride, Keoxifene hydrochloride, LY 156758, [6-Hydroxy-2-(4-hydroxyphenyl)benzo[b]thien-3-yl][4-[2-(1-piperidinyl)ethoxy]phenyl]methanone hydrochloride
Empirical Formula (Hill Notation):
C28H27NO4S · HCl
CAS Number:
Molecular Weight:
510.04
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing