
Rabeprazole Related Compound E

Synonym(s):
2-{{[4-(3-Methoxypropoxy)-3-methyl-2-pyridyl]methyl}thio}-1H-benzimidazole, Rabeprazole thioether
Empirical Formula (Hill Notation):
C18H21N3O2S
CAS Number:
Molecular Weight:
343.44
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2