
Prilocaine Related Compound B

Synonym(s):
(RS)-N-(Methylphenyl)-2-(propylamino)propanamide
Empirical Formula (Hill Notation):
C13H20N2O
CAS Number:
Molecular Weight:
220.31
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing