![Phosphatase, Alkaline from bovine intestinal mucosa ≥5,500 DEA units/mg protein](/deepweb/assets/sigmaaldrich/product/images/185/351/12833da1-90b8-43e0-9980-98edc47ee350/640/12833da1-90b8-43e0-9980-98edc47ee350.jpg)
Phosphatase, Alkaline from bovine intestinal mucosa
![Phosphatase, Alkaline from bovine intestinal mucosa ≥5,500 DEA units/mg protein](/deepweb/assets/sigmaaldrich/product/images/185/351/12833da1-90b8-43e0-9980-98edc47ee350/640/12833da1-90b8-43e0-9980-98edc47ee350.jpg)
Synonym(s):
Alkaline phosphatase, Orthophosphoric-monoester phosphohydrolase (alkaline optimum)
CAS Number:
Products
Brand
Biological Source
Physical Form
Product Line
Tissue
Quality Segment
Showing 1-6 of 6
Compare
Product Number
Description
Pricing