
N,N-Diethylformamide

Synonym(s):
N,N-Diethylformamide
Linear Formula:
HCON(C2H5)2
CAS Number:
Molecular Weight:
101.15
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing