
Narasin

Synonym(s):
(4S)-4-Methylsalinomycin
Empirical Formula (Hill Notation):
C43H72O11
CAS Number:
Molecular Weight:
765.03
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing