
Naltrindole hydrochloride

Synonym(s):
NTI hydrochloride
Empirical Formula (Hill Notation):
C26H26N2O3 · HCl
CAS Number:
Molecular Weight:
450.96
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing