
Methysergide maleate

Synonym(s):
Methysergide maleate salt
Empirical Formula (Hill Notation):
C21H27N3O2 · C4H4O4
CAS Number:
Molecular Weight:
469.53
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing