Skip to Content
Merck
vibrant-m

Methylprednisolone hydrogen succinate

vibrant-m
Synonym(s):
Methylprednisolone hemisuccinate, 11β,17,21-Trihydroxy-6α-methylpregna-1,4-diene-3,20-dione 21-succinate, Methylprednisolone hydrogen succinate
Empirical Formula (Hill Notation):
C26H34O8
CAS Number:
Molecular Weight:
474.54

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1