
Megestrol acetate for peak identification

Synonym(s):
Megestrol acetate, 17α-Acetoxy-6-methyl-4,6-pregnadiene-3,20-dione, 17α-Hydroxy-6-methyl-4,6-pregnadiene-3,20-dione 17-acetate, 17α-Hydroxy-6-methyl-6-dehydroprogesterone 17-acetate, Megestrol 17-acetate
Empirical Formula (Hill Notation):
C24H32O4
CAS Number:
Molecular Weight:
384.51
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing