
L-Methionine sulfoximine

Synonym(s):
L-S-(3-Amino-3-carboxypropyl)-S-methylsulfoximine
Linear Formula:
CH3S(O)(=NH)CH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
180.23
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Technique