![Klebsiella aerogenes (formerly Enterobacter aerogenes) WDCM 00175 Vitroids™ 130-300 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/images/320/629/b5ccb6ec-9bbd-450b-9069-7f3ceadcd577/640/b5ccb6ec-9bbd-450b-9069-7f3ceadcd577.jpg)
Klebsiella aerogenes (formerly Enterobacter aerogenes) WDCM 00175 Vitroids™
![Klebsiella aerogenes (formerly Enterobacter aerogenes) WDCM 00175 Vitroids™ 130-300 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/images/320/629/b5ccb6ec-9bbd-450b-9069-7f3ceadcd577/640/b5ccb6ec-9bbd-450b-9069-7f3ceadcd577.jpg)
Products
Brand
Manufacturer
Physical Form
Agency
Quality Segment
Application
Microorganism Suitability
Package Size
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing