Ibuprofen Tetraethylene Glycol Diester
Synonym(s):
Ibuprofen Tetraethylene Glycol Diester, ((oxybis(ethane-2,1-diyl))bis(oxy))bis(ethane-2,1-diyl) bis(2-(4-isobutylphenyl)propanoate)), 1,1′-[Oxybis(2,1-ethanediyloxy-2,1-ethanediyl)] bis[α-methyl-4-(2-methylpropyl)benzeneacetate]
Empirical Formula (Hill Notation):
C34H50O7
CAS Number:
Molecular Weight:
570.76
Products
Brand
Feature
Formula Weight
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2