Henderson Molybdenite
Synonym(s):
Molybdenum(IV) sulfide, Molybdenum disulfide
Empirical Formula (Hill Notation):
MoS2
CAS Number:
Molecular Weight:
160.07
Products
Formula Weight
Manufacturer
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing