
Ginkgolide C

Synonym(s):
1,7-Dihydroxy-ginkgolide A
Empirical Formula (Hill Notation):
C20H24O11
CAS Number:
Molecular Weight:
440.40
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing