
Flumazenil

Synonym(s):
Ro 15-1788, Ro15-1788, Flumazenil
Empirical Formula (Hill Notation):
C15H14FN3O3
CAS Number:
Molecular Weight:
303.29
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade