
Docetaxel for system suitability

Synonym(s):
Docetaxel trihydrate
Empirical Formula (Hill Notation):
C43H53NO14 · 3H2O
CAS Number:
Molecular Weight:
861.93
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing