Skip to Content
CD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dimethyl terephthalate
Synonym(s):
Dimethyl terephthalate
Linear Formula:
C
6
H
4
-1,4-(CO
2
CH
3
)
2
CAS Number:
120-61-6
Molecular Weight:
194.18
EC Number:
204-411-8
Products
Product Category
building blocks (2)
plasticizer (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Purum (1)
certified reference material (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
185124
Dimethyl terephthalate,
ReagentPlus
®
, ≥99%
Expand
View Pricing
8.08177
Dimethyl terephthalate,
for synthesis
Expand
View Pricing
86440
Dimethyl terephthalate,
purum, ≥99.0% (GC)
Expand
View Pricing
07038
Dimethyl terephthalate,
Trace
CERT
®
, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing