
Dihydrokaempferol

Synonym(s):
(+)-Aromadendrol, (+)-Dihydrokaempferol, (2R,3R)-3,4′,5,7-Tetrahydroxyflavanone, (2R,3R)-3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-one, Aromadendrin, Katuranin
Empirical Formula (Hill Notation):
C15H12O6
CAS Number:
Molecular Weight:
288.25
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing