
Dibrompropamidine diisetionate

Empirical Formula (Hill Notation):
C21H30Br2N4O10S2
CAS Number:
Molecular Weight:
722.42
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing